4-Brom-3-Chlorphenol ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
B5816-1G
B5816-5G
TCIAB5816-1GEA
444
DKK
TCIAB5816-1G
TCIAB5816-5G
4-Brom-3-Chlorphenol ≥98.0% (by GC, titration analysis)
4-Brom-3-Chlorphenol
Formel:
C₆H₄BrClO Molekylvægt: 207.45 g/mol |
MDL Number:
MFCD00070744 CAS-nummer: 13631-21-5 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 66.0 to 70.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...