4-([1,1'-Biphenyl]-4-yl)-6-chloro-2-phenylpyrimidine ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

B5802-1G B5802-5G
TCIAB5802-1GEA 501 DKK
TCIAB5802-1G TCIAB5802-5G
4-([1,1'-Biphenyl]-4-yl)-6-chloro-2-phenylpyrimidine ≥98.0% (by GC)
4-([1,1'-Biphenyl]-4-yl)-6-chloro-2-phenylpyrimidine
Formel: C₂₂H₁₅ClN₂
Molekylvægt: 342.83 g/mol
MDL Number: MFCD29477698
CAS-nummer: 1689538-58-6

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 144.0 to 148.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR