Bisindolylmaleimide I ≥97.0% (by HPLC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: GF 109203X

B5781-25MG B5781-5MG
TCIAB5781-25MGEA 3290 DKK
TCIAB5781-25MG TCIAB5781-5MG
Bisindolylmaleimide I ≥97.0% (by HPLC)
Bisindolylmaleimide I
Formel: C₂₅H₂₄N₄O₂
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00236428
CAS-nummer: 133052-90-1

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Melting point 268.0 to 272.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR