Bis(3-sulpho-N-succinimidyl) Suberate disodium salt ≥96.0% (by total nitrogen basis)
Leverandør: TCISamlet vurdering
Gennemsntilig vurdering
Synonymer:
1,8-Bis(2,5-dioxo-3-sulpho-1-pyrrolidinyl) Octanedioate disodium salt, BS3 Crosslinker disodium, Sodium 1,1'-(octanedioylbis(oxy))bis(2,5-dioxopyrrolidine-3-sulphonate), Suberic acid bis(3-sulpho-N-succinimidyl) ester disodium salt, sulpho-DSS 2Na, Octanedioic acid 1,8-Bis(2,5-dioxo-3-sulpho-1-pyrrolidinyl) ester disodium salt, Sodium 1,1'-[Octanedioylbis(oxy)]bis(2,5-dioxopyrrolidine-3-sulphonate)
Formel:
C₁₆H₁₈N₂Na₂O₁₄S₂ Molekylvægt: 572.42 g/mol Opbevaringstemperatur: Fryser |
MDL Number:
MFCD00054982 CAS-nummer: 127634-19-9 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (with Total Nitrogen) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...