4-Bromo-9-phenyl-9H-carbazole ≥97.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
B5767-1G
B5767-200MG
TCIAB5767-1GEA
1240
DKK
TCIAB5767-1G
TCIAB5767-200MG
4-Bromo-9-phenyl-9H-carbazole ≥97.0% (by GC)
4-Bromo-9-phenyl-9H-carbazole
Formel:
C₁₈H₁₂BrN |
MDL Number:
MFCD29067336 CAS-nummer: 1097884-37-1 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...