Bentranil ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

B5739-1G B5739-250MG
TCIAB5739-1GEA 2110 DKK
TCIAB5739-1G TCIAB5739-250MG
Bentranil ≥98.0% (by GC)
Bentranil
Formel: C₁₄H₉NO₂
Kogepunkt: 192°C
MDL Number: MFCD00043598
CAS-nummer: 1022-46-4

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 122.0 to 126.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR