N,N'-Bis(4-methoxyphenyl)benzidine ≥98.0% (by HPLC, total nitrogen)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: N,N'-Bis(4-methoxyphenyl)-[1,1'-biphenyl]-4,4'-diamine, N4,N4'-Bis(4-methoxyphenyl)-[1,1'-biphenyl]-4,4'-diamine

B5633-1G B5633-5G
TCIAB5633-1GEA 739 DKK
TCIAB5633-1G TCIAB5633-5G
N,N'-Bis(4-methoxyphenyl)benzidine ≥98.0% (by HPLC, total nitrogen)
N,N'-Bis(4-methoxyphenyl)benzidine
Formel: C₂₆H₂₄N₂O₂
MDL Number: MFCD32062860
CAS-nummer: 59131-00-9

Bestil nu

Specifikationer Testresultater

Appearance White to Amber to Dark purple powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 225.0 to 229.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR