Benidipine hydrochloride ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 3-[(3RS)-1-Benzylpiperidin-3-yl] 5-Methyl (4RS)-2,6-Dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate hydrochloride, KW‐3049

B5629-100MG B5629-1G
TCIAB5629-100MGEA 780 DKK
TCIAB5629-100MG TCIAB5629-1G
Benidipine hydrochloride ≥98.0% (by HPLC, titration analysis)
Benidipine hydrochloride
Formel: C₂₈H₃₁N₃O₆·HCl
Molekylvægt: 542.03 g/mol
Smeltepunkt: 200°C
Opbevaringstemperatur: Fryser
MDL Number: MFCD00906929
CAS-nummer: 91599-74-5
UN nummer: 2811
ADR: 6.1 III
Merck Index: 14,01043
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d832e5bbbddc43eadf3888ecd57515c12d826787&vid=821d68cc8fd08df8c1d22f7cd2eb3be1fd5f0686&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR