3-Bromo-9-(3,5-dimethylphenyl)-9H-carbazole ≥98.0% (by HPLC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

B5621-200MG B5621-1G
TCIAB5621-200MGEA 591 DKK
TCIAB5621-200MG TCIAB5621-1G
3-Bromo-9-(3,5-dimethylphenyl)-9H-carbazole ≥98.0% (by HPLC)
3-Bromo-9-(3,5-dimethylphenyl)-9H-carbazole
Formel: C₂₀H₁₆BrN
CAS-nummer: 1141017-77-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=28975b126592cabfa67e0dba9d99536a266447b5&vid=28975b126592cabfa67e0dba9d99536a266447b5&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 113.0 to 117.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR