3-Bromo-9-hexyl-9H-carbazole ≥99.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
B5608-1G
B5608-5G
TCIAB5608-1GEA
723
DKK
TCIAB5608-1G
TCIAB5608-5G
3-Bromo-9-hexyl-9H-carbazole ≥99.0% (by GC)
3-Bromo-9-hexyl-9H-carbazole
Formel:
C₁₈H₂₀BrN |
MDL Number:
MFCD30562084 CAS-nummer: 156972-74-6 |
|
Specifikationer Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 99.0 % |
Melting point | 47.0 to 51.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...