1-Butylpyridinium bis(trifluoromethanesulphonyl)imide ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
B5571-5G
B5571-25G
TCIAB5571-5GEA
632
DKK
TCIAB5571-5G
TCIAB5571-25G
1-Butylpyridinium bis(trifluoromethanesulphonyl)imide ≥98.0% (by HPLC, titration analysis)
1-Butylpyridinium bis(trifluoromethanesulphonyl)imide
Formel:
C₁₁H₁₄F₆N₂O₄S₂ Molekylvægt: 416.35 g/mol |
MDL Number:
MFCD16038143 CAS-nummer: 187863-42-9 |
|
Specifikationer Testresultater
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (HPLC) | min. 98.0 area% |
Purity (Ion exchange titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...