2-Brom-4-(trifluormethyl)pyridin ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
B5510-1G
B5510-5G
TCIAB5510-1GEA
362
DKK
TCIAB5510-1G
TCIAB5510-5G
2-Brom-4-(trifluormethyl)pyridin ≥98.0% (by GC)
2-Bromo-4-(trifluoromethyl)pyridine
Formel:
C₆H₃BrF₃N Kogepunkt: 162°C Smeltepunkt: -20°C Flammepunkt: 73°C |
MDL Number:
MFCD00153085 CAS-nummer: 175205-81-9 |
Specifikationer Testresultater
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...