tert-Butyldimethylsilanol ≥95.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

B5499-5ML B5499-25ML
TCIAB5499-5MLEA 698 DKK
TCIAB5499-5ML TCIAB5499-25ML
tert-Butyldimethylsilanol ≥95.0% (by GC)
tert-Butyldimethylsilanol
Formel: C₆H₁₆OSi
Kogepunkt: 139°C
Flammepunkt: 45°C
MDL Number: MFCD00010673
CAS-nummer: 18173-64-3
UN nummer: 1987
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=36d90459c936c0ca308bb5b258789fbedaf52e80&vid=36d90459c936c0ca308bb5b258789fbedaf52e80&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR