9-Benzyl-3-bromo-9H-carbazole ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

B5461-1G B5461-5G
TCIAB5461-1GEA 509 DKK
TCIAB5461-1G TCIAB5461-5G
9-Benzyl-3-bromo-9H-carbazole ≥98.0% (by GC)
9-Benzyl-3-bromo-9H-carbazole
Formel: C₁₉H₁₄BrN
MDL Number: MFCD00821698
CAS-nummer: 339576-55-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6358713744b2fe3db22679ed5eabc354acc0d631&vid=6358713744b2fe3db22679ed5eabc354acc0d631&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 116.0 to 120.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR