9-Benzyl-2-bromo-9H-carbazole ≥98.0% (by GC, total nitrogen)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
B5460-1G
B5460-5G
TCIAB5460-1GEA
928
DKK
TCIAB5460-1G
TCIAB5460-5G
9-Benzyl-2-bromo-9H-carbazole ≥98.0% (by GC, total nitrogen)
9-Benzyl-2-bromo-9H-carbazole
Formel:
C₁₉H₁₄BrN |
MDL Number:
MFCD30469064 CAS-nummer: 1401863-51-1 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 133.0 to 137.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...