2-Bromo-3-cyclohexylthiophene ≥95.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

B5420-1G B5420-5G
TCIAB5420-1GEA 1470 DKK
TCIAB5420-1G TCIAB5420-5G
2-Bromo-3-cyclohexylthiophene ≥95.0% (by GC)
2-Bromo-3-cyclohexylthiophene
Formel: C₁₀H₁₃BrS
Molekylvægt: 245.18 g/mol
Opbevaringstemperatur: Køleskab
CAS-nummer: 241477-71-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4046f964c9b864d0ac78deacfc2f8b6bba793e6&vid=c4046f964c9b864d0ac78deacfc2f8b6bba793e6&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR