7-Bromo-2,3-dihydrothieno[3,4-b][1,4]dioxine-5-carboxaldehyde ≥96.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

B5396-200MG B5396-1G
TCIAB5396-200MGEA 460 DKK
TCIAB5396-200MG TCIAB5396-1G
7-Bromo-2,3-dihydrothieno[3,4-b][1,4]dioxine-5-carboxaldehyde ≥96.0% (by GC)
7-Bromo-2,3-dihydrothieno[3,4-b][1,4]dioxine-5-carboxaldehyde
Formel: C₇H₅BrO₃S
MDL Number: MFCD08690306
CAS-nummer: 852054-42-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a272eb322022738315634d2c5d69a9c3ff212359&vid=a272eb322022738315634d2c5d69a9c3ff212359&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light gray to Light yellow powder to crystal
Purity (GC) min. 96.0 %
Melting point 147.0 to 152.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR