Butyl N-(3-Trifluoromethylphenyl)anthranilate ≥98.0% (by HPLC, total nitrogen)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Flufenamic acid Butyl ester, N-(3-Trifluoromethylphenyl)anthranilic acid Butyl ester, Butyl 2-[[3-(Trifluoromethyl)phenyl]amino]benzoate, Ufenamate, Butyl Flufenamate, Butyl 2-((3-(trifluoromethyl)phenyl)amino)benzoate, 2-[[3-(Trifluoromethyl)phenyl]amino]benzoic acid Butyl ester
B5391-200MG
B5391-1G
TCIAB5391-200MGEA
723
DKK
TCIAB5391-200MG
TCIAB5391-1G
Butyl N-(3-Trifluoromethylphenyl)anthranilate ≥98.0% (by HPLC, total nitrogen)
Butyl N-(3-Trifluoromethylphenyl)anthranilate
Formel:
C₁₈H₁₈F₃NO₂ Kogepunkt: 170°C |
MDL Number:
MFCD00866001 CAS-nummer: 67330-25-0 Merck Index: 14,04132 |
|
Specifikationer Testresultater
Appearance | White to Yellow to Green clear liquid |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...