1-Butoxy-2,3-difluoro-4-(trans-4-propylcyclohexyl)benzene ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

B5364-1G B5364-5G
TCIAB5364-1GEA 411 DKK
TCIAB5364-1G TCIAB5364-5G
1-Butoxy-2,3-difluoro-4-(trans-4-propylcyclohexyl)benzene ≥98.0% (by GC)
1-Butoxy-2,3-difluoro-4-(trans-4-propylcyclohexyl)benzene
Formel: C₁₉H₂₈F₂O
Molekylvægt: 310.43 g/mol
MDL Number: MFCD20489249
CAS-nummer: 208709-55-1

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR