Boc-Tyr-OBZL ≥98.0% (by HPLC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: (S)-Benzyl 2-((tert-butoxycarbonyl)amino)-3-(4-hydroxyphenyl)propanoate, N-Boc-L-tyrosine Benzyl ester, Boc-Tyr-OBzl, N-(tert-Butoxycarbonyl)-L-tyrosine Benzyl ester

B5343-1G B5343-5G
TCIAB5343-1GEA 403 DKK
TCIAB5343-1G TCIAB5343-5G
Boc-Tyr-OBZL ≥98.0% (by HPLC)
Boc-Tyr-OBZL
Formel: C₂₁H₂₅NO₅
MDL Number: MFCD00190840
CAS-nummer: 19391-35-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0d24d0e8cd819726eb288d3ef187fd49aa446201&vid=0d24d0e8cd819726eb288d3ef187fd49aa446201&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 95.0 %
Melting point 126.0 to 130.0 deg-C
Specific rotation [a]20/D -8.0 to -12.0 deg (C=1,MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR