2-(5-Bromo-2-methylbenzyl)-5-(4-fluorophenyl)thiophene ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

B5335-200MG B5335-1G
TCIAB5335-200MGEA 230 DKK
TCIAB5335-200MG TCIAB5335-1G
2-(5-Bromo-2-methylbenzyl)-5-(4-fluorophenyl)thiophene ≥98.0% (by GC)
2-(5-Bromo-2-methylbenzyl)-5-(4-fluorophenyl)thiophene
Formel: C₁₈H₁₄BrFS
MDL Number: MFCD21496340
CAS-nummer: 1030825-20-7

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 101.0 to 105.0 deg-C
melting range max. 5.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR