9-(2'-Bromo-4-biphenylyl)carbazole ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 9-(2'-Bromo-[1,1'-biphenyl]-4-yl)-9H-carbazole

B5312-200MG B5312-1G
TCIAB5312-200MGEA 493 DKK
TCIAB5312-200MG TCIAB5312-1G
9-(2'-Bromo-4-biphenylyl)carbazole ≥98.0% (by GC)
9-(2'-Bromo-4-biphenylyl)carbazole
Formel: C₂₄H₁₆BrN
Molekylvægt: 398.30 g/mol
MDL Number: MFCD31618099
CAS-nummer: 1215228-57-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=64962c84cc6be00298594328d8e0cc341bb0fddd&vid=64962c84cc6be00298594328d8e0cc341bb0fddd&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 104.0 to 108.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR