3-Brom-4-iodtoluen ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-Bromo-1-iodo-4-methylbenzene, 1-Bromo-2-iodo-5-methylbenzene

B5284-5G B5284-25G
TCIAB5284-5GEA 165 DKK
TCIAB5284-5G TCIAB5284-25G
3-Brom-4-iodtoluen ≥98.0% (by GC)
3-Brom-4-iodtoluen
Formel: C₇H₆BrI
Molekylvægt: 296.93 g/mol
Kogepunkt: 267°C
MDL Number: MFCD00079718
CAS-nummer: 71838-16-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=84e0e554c930c96317f449f788b9906229e37bed&vid=84e0e554c930c96317f449f788b9906229e37bed&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Brown clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR