3-Brom-2-methoxybenzoesyre ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
B5262-1G
B5262-5G
TCIAB5262-1GEA
181
DKK
TCIAB5262-1G
TCIAB5262-5G
3-Brom-2-methoxybenzoesyre ≥98.0% (by GC, titration analysis)
3-Brom-2-methoxybenzoesyre
Formel:
C₈H₇BrO₃ Molekylvægt: 231.05 g/mol |
MDL Number:
MFCD00045840 CAS-nummer: 101084-39-3 |
Specifikationer Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 120.0 to 124.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...