2-(4-Biphenylyl)indole ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 2-([1,1'-Biphenyl]-4-yl)-1H-indole

B5250-1G B5250-5G
TCIAB5250-1GEA 796 DKK
TCIAB5250-1G TCIAB5250-5G
2-(4-Biphenylyl)indole ≥98.0% (by GC)
2-(4-Biphenylyl)indole
Formel: C₂₀H₁₅N
Molekylvægt: 269.35 g/mol
MDL Number: MFCD00417190
CAS-nummer: 21470-37-1

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR