(S)-1-[3,5-Bis(trifluoromethyl)phenyl]ethanol ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
(S)-1-[3,5-Bis(trifluoromethyl)phenyl]ethyl alcohol, (S)-(-)-3,5-Bis(trifluoromethyl)-alpha-methylbenzyl alcohol, (1S)-(-)-1-[3,5-Bis(trifluoromethyl)phenyl]ethan-1-ol, (S)-1-(3,5-Bis(trifluoromethyl)phenyl)ethanol, (S)-3,5-Bis(trifluoromethyl)-alpha-methylbenzyl alcohol
B5244-1G
B5244-5G
TCIAB5244-1GEA
321
DKK
TCIAB5244-1G
TCIAB5244-5G
(S)-1-[3,5-Bis(trifluoromethyl)phenyl]ethanol ≥98.0% (by GC)
(S)-1-[3,5-Bis(trifluoromethyl)phenyl]ethanol
Formel:
C₁₀H₈F₆O Molekylvægt: 258.16 g/mol |
MDL Number:
MFCD03093011 CAS-nummer: 225920-05-8 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Optical purity (GC) | min. 98.0 ee% |
Melting point | 53.0 to 57.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...