1-Boc-1H-pyrazole-4-boronic acid pinacol ester ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-(1-tert-Butoxycarbonyl-4-pyrazolyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, N-Boc-pyrazole-4-boronic acid pinacol ester, tert-Butyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole-1-carboxylate, 1-(tert-Butoxycarbonyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole, 1-(tert-Butoxycarbonyl)pyrazolyl-4-boronic acid Pinacol ester, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole-1-carboxylic acid tert-butyl ester, 1-Boc-pyrazolyl-4-boronic acid Pinacol ester, 2-(1-Boc-4-pyrazolyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, tert-butyl 4-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole-1-carboxylate, 1-Boc-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole-1-carboxylic acid tert-butyl ester, tert-Butyl 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole-1-carboxylate

B5203-1G B5203-5G
TCIAB5203-1GEA 378 DKK
TCIAB5203-1G TCIAB5203-5G
1-Boc-1H-pyrazole-4-boronic acid pinacol ester ≥98.0% (by titrimetric analysis)
1-Boc-1H-pyrazole-4-boronic acid pinacol ester
Formel: C₁₄H₂₃N₂O₄B
Molekylvægt: 294.1544 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD05663873
CAS-nummer: 552846-17-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a6869990935fd62970dc8aa8fc1dc3b206d6ab14&vid=a6869990935fd62970dc8aa8fc1dc3b206d6ab14&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 85.0 to 89.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR