5-Bromo-2-ethynylpyridine ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

B5179-200MG B5179-1G
TCIAB5179-200MGEA 493 DKK
TCIAB5179-200MG TCIAB5179-1G
5-Bromo-2-ethynylpyridine ≥98.0% (by GC)
5-Bromo-2-ethynylpyridine
Formel: C₇H₄BrN
MDL Number: MFCD03095276
CAS-nummer: 111770-86-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=68a707e815ce5840fb50a532a97cce67ec29fb75&vid=68a707e815ce5840fb50a532a97cce67ec29fb75&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 83.0 to 87.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR