1-Boc-azetidine-3-carboxaldehyde ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 3-Formylazetidine-1-carboxylic acid tert-Butyl ester, 1-(tert-Butoxycarbonyl)azetidine-3-carboxaldehyde, tert-Butyl 3-Formylazetidine-1-carboxylate, 1-Boc-azetidine-3-carboxaldehyde

B5160-200MG B5160-1G
TCIAB5160-200MGEA 329 DKK
TCIAB5160-200MG TCIAB5160-1G
1-Boc-azetidine-3-carboxaldehyde ≥98.0% (by GC)
1-Boc-azetidine-3-carboxaldehyde
Formel: C₉H₁₅NO₃
Molekylvægt: 185.22 g/mol
MDL Number: MFCD06656142
CAS-nummer: 177947-96-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=97296c542c75b728f818261af113b7729bf0f817&vid=97296c542c75b728f818261af113b7729bf0f817&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR