3-Bromo-5-phenylpyridine ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

B5151-200MG B5151-1G
TCIAB5151-200MGEA 427 DKK
TCIAB5151-200MG TCIAB5151-1G
3-Bromo-5-phenylpyridine ≥98.0% (by GC, titration analysis)
3-Bromo-5-phenylpyridine
Formel: C₁₁H₈BrN
Kogepunkt: 101°C
MDL Number: MFCD04114223
CAS-nummer: 142137-17-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=121aab5d07597658afb0c26b8308706da2303812&vid=121aab5d07597658afb0c26b8308706da2303812&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 46.0 to 50.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR