2-[3,5-Bis(trifluoromethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-(3,5-Bis(trifluoromethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1,3-Bis(trifluoromethyl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene, 3,5-Bis(trifluoromethyl)phenylboronic acid Pinacol ester

B5095-1G B5095-5G
TCIAB5095-1GEA 230 DKK
TCIAB5095-1G TCIAB5095-5G
2-[3,5-Bis(trifluoromethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥98.0% (by GC, titration analysis)
2-[3,5-Bis(trifluoromethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formel: C₁₄H₁₅BF₆O₂
Molekylvægt: 340.07 g/mol
MDL Number: MFCD12405516
CAS-nummer: 69807-91-6

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 65.0 to 69.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR