8-Benzyltheophylline ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
B5078-25G
B5078-5G
TCIAB5078-25GEA
2900
DKK
TCIAB5078-25G
TCIAB5078-5G
8-Benzyltheophylline ≥98.0% (by HPLC, titration analysis)
8-Benzyltheophylline
Formel:
C₁₄H₁₄N₄O₂ Smeltepunkt: 288°C |
MDL Number:
MFCD00067027 CAS-nummer: 2879-15-4 EINECS: 220-725-8 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...