3-Bromo-4-methylbenzaldehyde ≥96.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

B5073-1G B5073-5G
TCIAB5073-1GEA 173 DKK
TCIAB5073-1G TCIAB5073-5G
3-Bromo-4-methylbenzaldehyde ≥96.0% (by GC)
3-Bromo-4-methylbenzaldehyde
Formel: C₈H₇BrO
Molekylvægt: 199.0500 g/mol
MDL Number: MFCD04971197
CAS-nummer: 36276-24-1

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 96.0 %
Melting point 47.0 to 51.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR