4-Benzyloxy-3-chlorophenylboronic acid (contains varying amounts of Anhydride)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

B5064-1G B5064-5G
TCIAB5064-1GEA 156 DKK
TCIAB5064-1G TCIAB5064-5G
4-Benzyloxy-3-chlorophenylboronic acid (contains varying amounts of Anhydride)
4-Benzyloxy-3-chlorophenylboronic acid
Formel: C₁₃H₁₂BClO₃
MDL Number: MFCD04115642
CAS-nummer: 845551-44-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8e242578c09af15233f42ec0d92ee7f5765c0cde&vid=8e242578c09af15233f42ec0d92ee7f5765c0cde&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) gt. 97.0 and le 108.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR