2-Bromo-9-(2-naphthyl)-9H-carbazole ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
2-Bromo-9-(naphthalen-2-yl)-9H-carbazole
B5051-200MG
B5051-1G
TCIAB5051-200MGEA
362
DKK
TCIAB5051-200MG
TCIAB5051-1G
2-Bromo-9-(2-naphthyl)-9H-carbazole ≥98.0% (by GC)
2-Bromo-9-(2-naphthyl)-9H-carbazole
Formel:
C₂₂H₁₄BrN |
MDL Number:
MFCD26127422 CAS-nummer: 1427316-53-7 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 141.0 to 145.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...