3-Bromo-9-(1-naphthyl)-9H-carbazole ≥97.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 3-Bromo-9-(naphthalen-1-yl)-9H-carbazole

B5050-200MG B5050-1G
TCIAB5050-200MGEA 189 DKK
TCIAB5050-200MG TCIAB5050-1G
3-Bromo-9-(1-naphthyl)-9H-carbazole ≥97.0% (by GC)
3-Bromo-9-(1-naphthyl)-9H-carbazole
Formel: C₂₂H₁₄BrN
MDL Number: MFCD26127423
CAS-nummer: 934545-83-2

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR