2-(2-Bromophenyl)pyridine ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
B5044-200MG
B5044-1G
TCIAB5044-200MGEA
501
DKK
TCIAB5044-200MG
TCIAB5044-1G
2-(2-Bromophenyl)pyridine ≥98.0% (by GC, titration analysis)
2-(2-Bromophenyl)pyridine
Formel:
C₁₁H₈BrN Kogepunkt: 139°C |
MDL Number:
MFCD15529755 CAS-nummer: 109306-86-7 |
|
Specifikationer Testresultater
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...