(4-Bromophenyl)phosphonic acid ≥98.0% (by HPLC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

B5015-200MG B5015-1G
TCIAB5015-200MGEA 632 DKK
TCIAB5015-200MG TCIAB5015-1G
(4-Bromophenyl)phosphonic acid ≥98.0% (by HPLC)
(4-Bromophenyl)phosphonic acid
Formel: C₆H₆BrO₃P
MDL Number: MFCD00159409
CAS-nummer: 16839-13-7

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 97.0 %
Melting point 200.0 to 204.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR