2-Butynyl toluene-4-sulphonate

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 2-Butynyl p-toluenesulphonate, But-2-yn-1-yl 4-methylbenzenesulphonate

B4995-1G B4995-5G
TCIAB4995-1GEA 788 DKK
TCIAB4995-1G TCIAB4995-5G
2-Butynyl toluene-4-sulphonate
2-Butynyl toluene-4-sulphonate
Formel: C₁₁H₁₂O₃S
Molekylvægt: 224.276 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00078395
CAS-nummer: 56563-37-2

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 47.0 to 51.0 deg-C

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR