1-Bromo-3-chloro-5-iodobenzene ≥97.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

B4981-1G B4981-5G
TCIAB4981-1GEA 485 DKK
TCIAB4981-1G TCIAB4981-5G
1-Bromo-3-chloro-5-iodobenzene ≥97.0% (by GC)
1-Bromo-3-chloro-5-iodobenzene
Formel: C₆H₃BrClI
Molekylvægt: 317.3500 g/mol
MDL Number: MFCD07778998
CAS-nummer: 13101-40-1

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 97.0 %
Melting point 84.0 to 88.4 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR