3-Brom-5-nitroanisol ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
1-Bromo-3-methoxy-5-nitrobenzene
B4973-1G
B4973-5G
TCIAB4973-1GEA
468
DKK
TCIAB4973-1G
TCIAB4973-5G
3-Brom-5-nitroanisol ≥98.0% (by GC)
3-Brom-5-nitroanisol
Formel:
C₇H₆BrNO₃ |
MDL Number:
MFCD04112752 CAS-nummer: 16618-67-0 |
Specifikationer Testresultater
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 86.0 to 90.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...