3-Bromo-6-chlorochromone ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
3-Bromo-6-chloro-4H-chromen-4-one
B4967-1G
B4967-5G
TCIAB4967-1GEA
468
DKK
TCIAB4967-1G
TCIAB4967-5G
3-Bromo-6-chlorochromone ≥98.0% (by GC)
3-Bromo-6-chlorochromone
Formel:
C₉H₄BrClO₂ Molekylvægt: 259.49 g/mol |
MDL Number:
MFCD02093123 CAS-nummer: 73220-38-9 |
|
Specifikationer Testresultater
Purity (GC) | min. 98.0 % |
Melting point | 129.0 to 133.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...