9,10-Bis[N-(m-tolyl)anilino]anthracene ≥98.0% (by HPLC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: N,N'-Diphenyl-N,N'-di(m-tolyl)-9,10-anthracenediamine, N,N'-Bis(3-methylphenyl)-N,N'-diphenyl-9,10-anthracenediamine, N9,N10-Diphenyl-N9,N10-di-m-tolylanthracene-9,10-diamine

B4965-200MG B4965-1G
TCIAB4965-200MGEA 362 DKK
TCIAB4965-200MG TCIAB4965-1G
9,10-Bis[N-(m-tolyl)anilino]anthracene ≥98.0% (by HPLC)
9,10-Bis[N-(m-tolyl)anilino]anthracene
Formel: C₄₀H₃₂N₂
Molekylvægt: 540.71 g/mol
MDL Number: MFCD12022466
CAS-nummer: 189263-81-8

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR