2,7-Bis[N,N-bis(4-methoxyphenyl)amino]-9,9-spirobi[9H-fluorene] ≥98.0% (by HPLC, total nitrogen)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
N,N,N',N'-Tetrakis(4-methoxyphenyl)-9,9'-spirobi[9H-fluorene]-2,7-diamine
B4959-200MG
B4959-1G
TCIAB4959-200MGEA
1090
DKK
TCIAB4959-200MG
TCIAB4959-1G
2,7-Bis[N,N-bis(4-methoxyphenyl)amino]-9,9-spirobi[9H-fluorene] ≥98.0% (by HPLC, total nitrogen)
2,7-Bis[N,N-bis(4-methoxyphenyl)amino]-9,9-spirobi[9H-fluorene]
Formel:
C₅₃H₄₂N₂O₄ |
CAS-nummer:
1138220-69-5 |
|
Specifikationer Testresultater
Appearance | White to Yellow to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 184.0 to 188.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...