4-(Biphenyl-4-yl)-6-(4-bromophenyl)-2-phenylpyrimidine ≥98.0% (by HPLC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 4-([1,1'-Biphenyl]-4-yl)-6-(4-bromophenyl)-2-phenylpyrimidine

B4953-1G B4953-5G
TCIAB4953-1GEA 714 DKK
TCIAB4953-1G TCIAB4953-5G
4-(Biphenyl-4-yl)-6-(4-bromophenyl)-2-phenylpyrimidine ≥98.0% (by HPLC)
4-(Biphenyl-4-yl)-6-(4-bromophenyl)-2-phenylpyrimidine
Formel: C₂₈H₁₉BrN₂
Molekylvægt: 463.38 g/mol
MDL Number: MFCD28975104
CAS-nummer: 1421599-34-9

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 95.0 %
Melting point 189.0 to 193.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR