Z-Glu-OtBu ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
1-tert-Butyl N-Carbobenzoxy-L-glutamate, N-Cbz-L-glutamic acid 1-tert-Butyl ester, 1-tert-Butyl N-Cbz-L-glutamate, N-Carbobenzoxy-L-glutamic acid 1-tert-Butyl ester, Z-Glu-OtBu, 1-tert-Butyl N-Benzyloxycarbonyl-L-glutamate, N-Benzyloxycarbonyl-L-glutamic acid 1-tert-butyl ester, N-Benzyloxycarbonyl-L-glutamic acid 1-tert-Butyl ester
B4912-5G
B4912-1G
TCIAB4912-5GEA
608
DKK
TCIAB4912-5G
TCIAB4912-1G
Z-Glu-OtBu ≥98.0% (by HPLC, titration analysis)
Z-Glu-OtBu
Formel:
C₁₇H₂₃NO₆ |
MDL Number:
MFCD00077016 CAS-nummer: 5891-45-2 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 81.0 to 85.0 deg-C |
Specific rotation [a]20/D | -24.0 to -28.0 deg (C=1,MeOH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...