3,3'-Bi[1,4]benzoxazino[2,3,4-kl]phenoxazine

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: HN-D2

B4907-200MG B4907-1G
TCIAB4907-200MGEA 1470 DKK
TCIAB4907-200MG TCIAB4907-1G
3,3'-Bi[1,4]benzoxazino[2,3,4-kl]phenoxazine
3,3'-Bi[1,4]benzoxazino[2,3,4-kl]phenoxazine
Formel: C₃₆H₂₀N₂O₄
Smeltepunkt: 340°C
CAS-nummer: 1395881-58-9

Bestil nu

Specifikationer Testresultater

Appearance Light orange to Yellow to Green powder to crystal
Elemental analysis (Nitrogen) 4.90 to 5.40 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR