4-Bromo-4'-[di(p-tolyl)amino]stilbene ≥97.0% (by HPLC, total nitrogen)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 4-(4-Bromostyryl)-N,N-di(p-tolyl)aniline, 4-(4-Bromostyryl)-N,N-di-p-tolylaniline, 4-(4-Bromostyryl)-4',4''-dimethyltriphenylamine

B4906-200MG B4906-1G
TCIAB4906-200MGEA 394 DKK
TCIAB4906-200MG TCIAB4906-1G
4-Bromo-4'-[di(p-tolyl)amino]stilbene ≥97.0% (by HPLC, total nitrogen)
4-Bromo-4'-[di(p-tolyl)amino]stilbene
Formel: C₂₈H₂₄BrN
MDL Number: MFCD28975103
CAS-nummer: 101186-77-0

Bestil nu

Specifikationer Testresultater

Appearance White to Yellow to Green powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (with Total Nitrogen) min. 97.0 %
Melting point 155.0 to 159.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR