1,4-Bis(2-hydroxyethyl)piperazine ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
Synonymer: 2,2'-(Piperazine-1,4-diyl)diethanol
B4903-5G B4903-25G
TCIAB4903-5GEA 148 DKK
TCIAB4903-5G TCIAB4903-25G
1,4-Bis(2-hydroxyethyl)piperazine ≥98.0% (by GC, titration analysis)
1,4-Bis(2-hydroxyethyl)piperazine
Formel: C₈H₁₈N₂O₂
Molekylvægt: 174.24 g/mol
Kogepunkt: 278°C
MDL Number: MFCD00006157
CAS-nummer: 122-96-3
EINECS: 204-586-0
Bestil nu
Specifikationer Testresultater
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 136.0 to 140.0 deg-C