9-(2-Biphenylyl)-10-bromoanthracene ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
9-([1,1'-Biphenyl]-2-yl)-10-bromoanthracene
B4895-1G
B4895-5G
TCIAB4895-1GEA
542
DKK
TCIAB4895-1G
TCIAB4895-5G
9-(2-Biphenylyl)-10-bromoanthracene ≥98.0% (by GC)
9-(2-Biphenylyl)-10-bromoanthracene
Formel:
C₂₆H₁₇Br |
MDL Number:
MFCD28138083 CAS-nummer: 400607-16-1 |
Specifikationer Testresultater
Appearance | Light yellow to Amber to Dark green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 146.0 to 150.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...